Assay Details
Target Gene Details
Entrez Gene ID: | 228677 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 3 |
Gene Aliases: |
C130053K05Rik |
Location: |
Chr.2:139493913-139637674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc3 | NM_175467.3 | NP_780676.1 | ||
| AK048374.1 | BAC33316.1 | |||
| AK054240.1 | BAC35701.1 | |||
| AK078679.1 | BAC37356.1 | |||
| AK078686.1 | BAC37359.1 | |||
| BC094496.1 | AAH94496.1 |
More Information
Set Membership: |
|

Genomic Map