Assay Details
Target Gene Details
Entrez Gene ID: | 20568 |
Gene Name: | secretory leukocyte peptidase inhibitor |
Gene Aliases: |
- |
Location: |
Chr.2:164354070-164356507 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slpi | NM_011414.3 | 3 | 566 | NP_035544.1 |
| AK010170.1 | 3 | 353 | ||
| BC028509.1 | 3 | 566 | AAH28509.1 | |
| U88093.1 | 3 | 343 | AAC53140.1 | |
| U94341.1 | 3 | 344 | AAC53394.1 |
More Information
Set Membership: |
|

Genomic Map