Assay Details
Target Gene Details
Entrez Gene ID: | 58231 |
Gene Name: | serine/threonine kinase 4 |
Gene Aliases: |
AI447339, AU020804, Kas-2, Mst1, Ysk3 |
Location: |
Chr.2:164074109-164155524 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk4 | NM_021420.3 | NP_067395.1 | ||
| AF271360.1 | AAF75789.1 | |||
| AK028838.1 | BAC26147.1 | |||
| BC054521.1 | AAH54521.1 |
More Information
Set Membership: |
|

Genomic Map