Assay Details
Target Gene Details
Entrez Gene ID: | 320106 |
Gene Name: | solute carrier family 38, member 11 |
Gene Aliases: |
9330158F14Rik |
Location: |
Chr.2:65315799-65364394 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc38a11 | NM_177074.2 | NP_796048.2 | ||
| AK034126.1 | 6 | 1606 | BAC28596.1 | |
| AK041015.1 | 5 | 1429 | BAC30781.1 | |
| AK043610.1 | ||||
| AK139966.1 | BAE24200.1 |
More Information
Set Membership: |
|

Genomic Map