Assay Details
Target Gene Details
Entrez Gene ID: | 22325 |
Gene Name: | vav 2 oncogene |
Gene Aliases: |
2810040F13Rik, AI847175 |
Location: |
Chr.2:27262104-27427608 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vav2 | NM_009500.1 | NP_033526.1 | ||
| BC053060.1 | AAH53060.1 | |||
| U37017.1 | AAC52761.1 |

Genomic Map