Assay Details
Target Gene Details
Entrez Gene ID: | 24128 |
Gene Name: | 5'-3' exoribonuclease 2 |
Gene Aliases: |
- |
Location: |
Chr.2:147012631-147078002 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrn2 | NM_011917.2 | 15 | 1410 | NP_036047.2 |
| AK004800.1 | 15 | 1385 | BAB23573.1 | |
| AK011711.1 | 15 | 1408 | ||
| AK031247.1 | 15 | 1453 | BAC27318.1 | |
| AK053643.1 | 15 | 1403 | BAC35458.1 | |
| AK167019.1 | 15 | 1430 | BAE39193.1 | |
| AK168037.1 | 15 | 1456 | BAE40020.1 | |
| BC004028.1 | 7 | 581 | AAH04028.1 | |
| BC054743.1 | 15 | 1410 | AAH54743.1 | |
| D38517.1 | 15 | 1348 | BAA07524.1 |
More Information
Set Membership: |
|

Genomic Map