Assay Details
Target Gene Details
Entrez Gene ID: | 52837 |
Gene Name: | thioredoxin-related transmembrane protein 4 |
Gene Aliases: |
2810417D04Rik, 4930500L08Rik, AI843224, AW046784, D2Bwg1356e, Txndc13, mKIAA1162 |
Location: |
Chr.2:134594501-134644121 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmx4 | NM_029148.1 | NP_083424.1 | ||
| AK015667.1 | ||||
| AK030696.1 | BAC27085.1 | |||
| AK033111.1 | ||||
| AK040779.1 | ||||
| AK053897.1 | ||||
| AK129301.1 | BAC98111.1 | |||
| AK147465.1 | BAE27931.1 | |||
| BC050918.1 | AAH50918.1 | |||
| BC054558.1 | ||||
| BC138168.1 | ||||
| BC138169.1 |

Genomic Map