Assay Details
Target Gene Details
Entrez Gene ID: | 76080 |
Gene Name: | tocopherol (alpha) transfer protein-like |
Gene Aliases: |
3110080A02Rik, 5830472M02Rik |
Location: |
Chr.2:163602281-163619013 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttpal | NM_029512.2 | 3 | 619 | NP_083788.2 |
| NM_181734.3 | 3 | 616 | NP_859423.2 | |
| AK018052.1 | ||||
| AK033600.1 | 3 | 565 | BAC28381.1 | |
| AK039450.1 | 3 | 617 | BAC30352.1 | |
| AK043791.1 | 3 | 586 | BAC31654.1 | |
| AK052794.1 | 3 | 582 | ||
| AK081680.1 | 3 | 579 | BAC38289.1 | |
| AK156641.1 | 3 | 582 | BAE33788.1 | |
| BC052389.1 | 3 | 575 | ||
| BY717985.1 | ||||
| CK640285.1 | 2 | 252 |
More Information
Set Membership: |
|

Genomic Map