Assay Details
Target Gene Details
Entrez Gene ID: | 21925 |
Gene Name: | troponin C2, fast |
Gene Aliases: |
Tncs |
Location: |
Chr.2:164777161-164779734 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnnc2 | NM_009394.2 | 6 | 570 | NP_033420.1 |
| AK133548.1 | 6 | 566 | BAE21718.1 | |
| BC024390.1 | 6 | 569 | AAH24390.1 |
More Information
Set Membership: |
|

Genomic Map