Assay Details
Target Gene Details
Entrez Gene ID: | 228012 |
Gene Name: | tousled-like kinase 1 |
Gene Aliases: |
4930545J15Rik |
Location: |
Chr.2:70712407-70825480 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlk1 | NM_172664.3 | 11 | 1263 | NP_766252.2 |
| AK029773.1 | 10 | 989 | BAC26610.1 | |
| AK172897.1 | 5 | 527 | BAD32175.1 | |
| BC033931.1 | 9 | 825 | ||
| BC051641.1 | 11 | 1263 | AAH51641.1 |
More Information
Set Membership: |
|

Genomic Map