Assay Details
Target Gene Details
Entrez Gene ID: | 228012 |
Gene Name: | tousled-like kinase 1 |
Gene Aliases: |
4930545J15Rik |
Location: |
Chr.2:70712407-70825480 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlk1 | NM_172664.3 | 6 | 649 | NP_766252.2 |
| AK029773.1 | 5 | 375 | BAC26610.1 | |
| AK045308.1 | 7 | 466 | ||
| BC033931.1 | 4 | 211 | ||
| BC051641.1 | 6 | 649 | AAH51641.1 |
More Information
Set Membership: |
|

Genomic Map