Assay Details
Target Gene Details
Entrez Gene ID: | 11722 |
Gene Name: | amylase 1, salivary |
Gene Aliases: |
Amy-1, Amy1a, C030014B17Rik |
Location: |
Chr.3:113555952-113577750 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amy1 | NM_001110505.1 | NP_001103975.1 | ||
| BC009121.1 | AAH09121.1 | |||
| V00717.1 | CAA24097.1 | |||
| V00720.1 | CAA24100.1 |
More Information
Set Membership: |
|

Genomic Map