Assay Details
Target Gene Details
Entrez Gene ID: | 66958 |
Gene Name: | thioredoxin-related transmembrane protein 2 |
Gene Aliases: |
2310042M24Rik, AA589631, Txndc14 |
Location: |
Chr.2:84671311-84678828 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmx2 | NM_001290751.1 | NP_001277680.1 | ||
| NM_025868.4 | NP_080144.1 | |||
| AK009759.1 | BAB26483.1 | |||
| AK075750.1 | BAC35930.1 | |||
| AK145245.1 | BAE26323.1 | |||
| AK170587.1 | BAE41896.1 | |||
| BC013544.1 | AAH13544.1 | |||
| BC110994.1 | AAI10995.1 |
More Information
Set Membership: |
|

Genomic Map