Assay Details
Target Gene Details
Entrez Gene ID: | 72147 |
Gene Name: | zinc finger and BTB domain containing 46 |
Gene Aliases: |
2610019F01Rik, 4933406L05Rik, BZEL, Btbd4 |
Location: |
Chr.2:181371261-181492982 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb46 | NM_027656.2 | 3 | 1347 | NP_081932.1 |
| AK016700.1 | 3 | 712 | BAB30386.1 | |
| AK038975.1 | 2 | 228 | BAC30190.1 |
More Information
Set Membership: |
|

Genomic Map