Assay Details
Target Gene Details
Entrez Gene ID: | 277353 |
Gene Name: | transcription factor-like 5 (basic helix-loop-helix) |
Gene Aliases: |
AV260129, Figlb |
Location: |
Chr.2:180621956-180642691 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcfl5 | NM_178254.3 | NP_839985.2 | ||
| AY234363.1 | AAO75014.1 | |||
| BC108399.1 | AAI08400.1 |
More Information
Set Membership: |
|

Genomic Map