Assay Details
Target Gene Details
Entrez Gene ID: | 338348 |
Gene Name: | tetratricopeptide repeat domain 16 |
Gene Aliases: |
1200002K10Rik |
Location: |
Chr.2:32757026-32775663 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc16 | NM_001290563.1 | NP_001277492.1 | ||
| NM_001290564.1 | NP_001277493.1 | |||
| NM_001290566.1 | NP_001277495.1 | |||
| NM_001290567.1 | NP_001277496.1 | |||
| NM_177384.3 | NP_796358.2 | |||
| AK028040.1 | BAC25714.1 | |||
| AK089384.1 | BAC40863.1 | |||
| AK133273.1 | BAE21588.1 | |||
| AK160435.1 | BAE35787.1 | |||
| BC140991.1 | ||||
| BC145064.1 |

Genomic Map