Assay Details
Target Gene Details
Entrez Gene ID: | 67701 |
Gene Name: | WAP four-disulfide core domain 2 |
Gene Aliases: |
1600023A02Rik, HE4, WAP5 |
Location: |
Chr.2:164562563-164568510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc2 | NM_026323.2 | NP_080599.1 | ||
| AF334269.1 | AAL73189.1 | |||
| AK005519.1 | BAB24094.1 | |||
| BC099427.1 | AAH99427.1 |
More Information
Set Membership: |
|

Genomic Map