Assay Details
Target Gene Details
Entrez Gene ID: | 229211 |
Gene Name: | acyl-Coenzyme A dehydrogenase family, member 9 |
Gene Aliases: |
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002 |
Location: |
Chr.3:36065984-36092857 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acad9 | NM_172678.3 | 3 | 410 | NP_766266.3 |
| AK028571.1 | 3 | 338 | ||
| AK031820.1 | 3 | 342 | BAC27565.1 | |
| AK075984.1 | 3 | 329 | BAC36096.1 | |
| AK145423.1 | 3 | 318 | BAE26429.1 | |
| AK189876.1 | 1 | 85 | ||
| AK192710.1 | 2 | 72 | ||
| AK195232.1 | 2 | 91 | ||
| AK198686.1 | 2 | 72 | ||
| AK200479.1 | 2 | 72 | ||
| AK205159.1 | 2 | 72 | ||
| AK216829.1 | 2 | 72 | ||
| BC032213.1 | 3 | 351 | AAH32213.1 | |
| BC033277.1 | 3 | 331 | AAH33277.1 |
More Information
Set Membership: |
|

Genomic Map