Assay Details
Target Gene Details
Entrez Gene ID: | 229211 |
Gene Name: | acyl-Coenzyme A dehydrogenase family, member 9 |
Gene Aliases: |
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002 |
Location: |
Chr.3:36065984-36092857 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acad9 | NM_172678.3 | NP_766266.3 | ||
| AK028571.1 | ||||
| AK031820.1 | BAC27565.1 | |||
| AK075984.1 | BAC36096.1 | |||
| AK145423.1 | BAE26429.1 | |||
| AK178887.1 | ||||
| AK179129.1 | ||||
| AK203069.1 | ||||
| AK206153.1 | ||||
| AK208945.1 | ||||
| AK215567.1 | ||||
| AK217282.1 | ||||
| BC032213.1 | AAH32213.1 | |||
| BC033277.1 | AAH33277.1 |
More Information
Set Membership: |
|

Genomic Map