Assay Details
Target Gene Details
Entrez Gene ID: | 109676 |
Gene Name: | ankyrin 2, brain |
Gene Aliases: |
AI835472, AW491075, Ank-2 |
Location: |
Chr.3:126921607-127499870 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank2 | NM_178655.3 | NP_848770.2 | ||
| AK032060.1 | BAC27676.1 | |||
| AK036018.1 | ||||
| AK044093.1 | ||||
| AK044634.1 | BAC32012.1 | |||
| AK082352.1 | ||||
| BC059251.1 | AAH59251.1 |
More Information
Set Membership: |
|

Genomic Map