Assay Details
Target Gene Details
Entrez Gene ID: | 209351 |
Gene Name: | WAP four-disulfide core domain 6A |
Gene Aliases: |
Gm122 |
Location: |
Chr.2:164579519-164585845 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc6a | NM_001033240.4 | NP_001028412.1 | ||
NM_001177848.1 | NP_001171319.1 | |||
AK136589.1 | BAE23064.1 | |||
BC147597.1 | ||||
BC147615.1 | ||||
BM933122.1 |
