Assay Details
Target Gene Details
Entrez Gene ID: | 433502 |
Gene Name: | WAP four-disulfide core domain 6B |
Gene Aliases: |
WAP6b, Wfdc6 |
Location: |
Chr.2:164613578-164618213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc6b | NM_001012725.2 | NP_001012743.2 | ||
AY541526.1 | AAT07038.1 | |||
BC141216.1 |
