Assay Details
Target Gene Details
Entrez Gene ID: | 545554 |
Gene Name: | ankyrin repeat domain 34A |
Gene Aliases: |
AW060394, Ankrd34 |
Location: |
Chr.3:96594826-96600126 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd34a | NM_001024851.3 | NP_001020022.2 | ||
| BC052924.1 | AAH52924.1 | |||
| BY125165.1 |

Genomic Map