Assay Details
Target Gene Details
Entrez Gene ID: | 22030 |
Gene Name: | TNF receptor-associated factor 2 |
Gene Aliases: |
AI325259 |
Location: |
Chr.2:25517982-25546940 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf2 | NM_001290413.1 | 2 | 192 | NP_001277342.1 |
| NM_009422.3 | 2 | 192 | NP_033448.2 | |
| AF027570.1 | 1 | 83 | AAC53545.1 | |
| AK038136.1 | 2 | 141 | BAC29937.1 | |
| AK052934.1 | 2 | 141 | BAC35209.1 | |
| AK151653.1 | 2 | 130 | BAE30582.1 | |
| AK152174.1 | 2 | 160 | BAE31006.1 | |
| AK159581.1 | 2 | 162 | BAE35201.1 | |
| BB841262.1 | 2 | 192 | ||
| BC003801.1 | 1 | 118 | AAH03801.1 | |
| L35303.1 | 2 | 129 | AAC37662.1 |
More Information
Set Membership: |
|

Genomic Map