Assay Details
Target Gene Details
Entrez Gene ID: | 22030 |
Gene Name: | TNF receptor-associated factor 2 |
Gene Aliases: |
AI325259 |
Location: |
Chr.2:25517982-25546940 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf2 | NM_001290413.1 | NP_001277342.1 | ||
| NM_009422.3 | NP_033448.2 | |||
| AF027570.1 | AAC53545.1 | |||
| AK038136.1 | BAC29937.1 | |||
| AK052934.1 | BAC35209.1 | |||
| AK151653.1 | BAE30582.1 | |||
| AK152174.1 | BAE31006.1 | |||
| AK159581.1 | BAE35201.1 | |||
| AK184085.1 | ||||
| AK184736.1 | ||||
| BC003801.1 | AAH03801.1 | |||
| L35303.1 | AAC37662.1 |
More Information
Set Membership: |
|

Genomic Map