Assay Details
Target Gene Details
Entrez Gene ID: | 22030 |
Gene Name: | TNF receptor-associated factor 2 |
Gene Aliases: |
AI325259 |
Location: |
Chr.2:25517982-25546940 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf2 | NM_001290413.1 | 5 | 576 | NP_001277342.1 |
| NM_009422.3 | 5 | 555 | NP_033448.2 | |
| AF027570.1 | 4 | 467 | AAC53545.1 | |
| AK038136.1 | 5 | 504 | BAC29937.1 | |
| AK052934.1 | 5 | 504 | BAC35209.1 | |
| AK151653.1 | 5 | 493 | BAE30582.1 | |
| AK152174.1 | 5 | 523 | BAE31006.1 | |
| AK159581.1 | 5 | 525 | BAE35201.1 | |
| AK184085.1 | ||||
| AK184736.1 | ||||
| BC003801.1 | 4 | 481 | AAH03801.1 | |
| L35303.1 | 5 | 492 | AAC37662.1 |
More Information
Set Membership: |
|

Genomic Map