Assay Details
Target Gene Details
Entrez Gene ID: | 629754 |
Gene Name: | WAP four-disulfide core domain 9 |
Gene Aliases: |
OTTMUSG00000001088 |
Location: |
Chr.2:164649625-164657094 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc9 | NM_001160414.1 | NP_001153886.1 | ||
| AK136630.1 | BAE23078.1 | |||
| AV381411.2 |
More Information
Set Membership: |
|

Genomic Map