Assay Details
Target Gene Details
Entrez Gene ID: | 73668 |
Gene Name: | tetratricopeptide repeat domain 21B |
Gene Aliases: |
2410066K11Rik, Thm1, aln, mKIAA1992 |
Location: |
Chr.2:66184327-66256675 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc21b | NM_001047604.2 | 14 | 1874 | NP_001041069.1 |
| NM_001290669.1 | 14 | 1874 | NP_001277598.1 | |
| AK147810.1 | 7 | 957 | BAE28153.1 | |
| AK164723.1 | 4 | 501 | BAE37891.1 | |
| DQ011164.1 | 14 | 1874 |

Genomic Map