Assay Details
Target Gene Details
Entrez Gene ID: | 100502700 |
Gene Name: | expressed sequence AV039307 |
Gene Aliases: |
- |
Location: |
Chr.2:120850656-120866690 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AV039307 | NR_038349.1 | 4 | 1020 | |
| NR_038350.1 | 4 | 558 | ||
| AK006022.1 | 4 | 510 | BAB24370.1 | |
| AK015969.1 | 4 | 960 | ||
| AV205317.1 | 1 | 172 | ||
| AV269478.1 | 1 | 97 |
Target Gene Details
Entrez Gene ID: | 22222 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 1 |
Gene Aliases: |
AI504731 |
Location: |
Chr.2:120860272-120970729 on Build GRCm38 |
Assay Gene Location: | Within Exon 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubr1 | NM_009461.2 | 45 | 5018 | NP_033487.2 |
| AF061555.1 | 45 | 5019 | AAC40165.1 | |
| AK078173.1 | 5 | 322 | BAC37160.1 |
More Information
Set Membership: |
|

Genomic Map