Assay Details
Target Gene Details
Entrez Gene ID: | 13722 |
Gene Name: | aminoacyl tRNA synthetase complex-interacting multifunctional protein 1 |
Gene Aliases: |
9830137A06Rik, AIMP1/p43, EMAPII, Emap2, Scye1, p43 |
Location: |
Chr.3:132660498-132683879 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aimp1 | NM_007926.2 | NP_031952.2 | ||
| AK036573.1 | ||||
| AK050571.1 | ||||
| AK087932.1 | BAC40045.1 | |||
| AK133859.1 | BAE21893.1 | |||
| AK160506.1 | BAE35830.1 | |||
| BC002054.1 | AAH02054.1 | |||
| BY256548.1 | ||||
| U10118.1 | AAA62203.1 |
More Information
Set Membership: |
|

Genomic Map