Assay Details
Target Gene Details
Entrez Gene ID: | 241494 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
B830010L13Rik, C130013B13Rik, Zfp533, Znf385b |
Location: |
Chr.2:77410622-77817706 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp385b | NM_001113399.1 | NP_001106870.1 | ||
| NM_001113400.1 | 1 | 89 | NP_001106871.1 | |
| NM_178723.6 | NP_848838.2 | |||
| AK046798.1 | BAC32874.1 | |||
| AK047859.1 | ||||
| AK050293.1 | 1 | 90 | BAC34170.1 | |
| AK078551.1 | ||||
| AK079322.1 | ||||
| AK149352.1 | BAE28828.1 | |||
| BC048543.1 | AAH48543.1 | |||
| BC132352.1 | ||||
| BC138239.1 |

Genomic Map