Assay Details
Target Gene Details
Entrez Gene ID: | 241494 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
B830010L13Rik, C130013B13Rik, Zfp533, Znf385b |
Location: |
Chr.2:77410622-77817706 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp385b | NM_001113399.1 | 5 | 755 | NP_001106870.1 |
| NM_001113400.1 | 5 | 777 | NP_001106871.1 | |
| NM_178723.6 | 7 | 1287 | NP_848838.2 | |
| AK046798.1 | 7 | 1288 | BAC32874.1 | |
| AK050293.1 | 5 | 778 | BAC34170.1 | |
| AK078551.1 | 5 | 731 | ||
| AK079322.1 | 7 | 1172 | ||
| AK149352.1 | 7 | 1268 | BAE28828.1 | |
| BC132352.1 | 5 | 866 | ||
| BC138239.1 | 5 | 866 |

Genomic Map