Assay Details
Target Gene Details
Entrez Gene ID: | 26377 |
Gene Name: | dual adaptor for phosphotyrosine and 3-phosphoinositides 1 |
Gene Aliases: |
Bam32 |
Location: |
Chr.3:137930998-137981576 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dapp1 | NM_011932.2 | 1 | 125 | NP_036062.1 |
| AF163255.1 | 1 | 79 | AAD49698.1 | |
| AF186023.1 | 1 | 100 | AAF14579.1 | |
| AK079794.1 | 1 | 113 | ||
| AK145745.1 | 1 | 91 | BAE26622.1 | |
| AK146180.1 | 1 | 94 | BAE26958.1 | |
| AK169584.1 | 1 | 113 | BAE41242.1 | |
| BC014759.1 | 1 | 121 | AAH14759.1 |

Genomic Map