Assay Details
Target Gene Details
Entrez Gene ID: | 208188 |
Gene Name: | growth hormone secretagogue receptor |
Gene Aliases: |
C530020I22Rik |
Location: |
Chr.3:27371351-27378010 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ghsr | NM_177330.4 | NP_796304.1 | ||
| AK049671.1 | BAC33866.1 |
More Information
Set Membership: |
|

Genomic Map