Assay Details
Target Gene Details
Entrez Gene ID: | 171388 |
Gene Name: | BCL2/adenovirus E1B 19kD interacting protein like |
Gene Aliases: |
1700128A13Rik, BNIP-S, BNIPL1, BNIPL2, PP73, PP753 |
Location: |
Chr.3:95241293-95251193 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bnipl | NM_001168356.1 | 4 | 612 | NP_001161828.1 |
| NM_134253.2 | 5 | 708 | NP_599014.2 | |
| AK142472.1 | 4 | 469 | BAE25078.1 | |
| BC005659.1 | 5 | 617 | AAH05659.1 |

Genomic Map