Assay Details
Target Gene Details
Entrez Gene ID: | 368202 |
Gene Name: | protease, serine 48 |
Gene Aliases: |
Esspl, Gm1019 |
Location: |
Chr.3:85973843-86006002 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss48 | NM_001001650.1 | NP_001001650.1 | ||
| BC116378.1 | ||||
| BC116379.1 |
Target Gene Details
Entrez Gene ID: | 27059 |
Gene Name: | SH3 domain protein D19 |
Gene Aliases: |
AW011754, Kryn |
Location: |
Chr.3:85970956-86130522 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3d19 | AK180019.1 |
More Information
Set Membership: |
|

Genomic Map