Assay Details
Target Gene Details
Entrez Gene ID: | 14866 |
Gene Name: | glutathione S-transferase, mu 5 |
Gene Aliases: |
- |
Location: |
Chr.3:107895854-107898685 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstm5 | NM_010360.2 | NP_034490.1 | ||
| AK002750.1 | BAB22327.1 | |||
| AK019317.1 | BAB31661.1 | |||
| AK028098.1 | BAC25746.1 | |||
| BC008206.1 | AAH08206.1 | |||
| U24428.1 | AAB04096.1 |
More Information
Set Membership: |
|

Genomic Map