Assay Details
Target Gene Details
Entrez Gene ID: | 70285 |
Gene Name: | ribosome production factor 1 homolog (S. cerevisiae) |
Gene Aliases: |
2210420E24Rik, 2310066N05Rik, Bxdc5 |
Location: |
Chr.3:146506344-146521431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpf1 | NM_027332.3 | NP_081608.1 | ||
NM_027371.3 | NP_081647.2 | |||
AK008988.1 | ||||
AK010079.1 | ||||
AK162073.1 | ||||
AK171035.1 | ||||
AK171079.1 | BAE42233.1 | |||
BC056165.1 | ||||
BC089369.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
