Assay Details
Target Gene Details
Entrez Gene ID: | 56811 |
Gene Name: | dickkopf homolog 2 (Xenopus laevis) |
Gene Aliases: |
- |
Location: |
Chr.3:132085242-132180304 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dkk2 | NM_020265.4 | 3 | 1151 | NP_064661.2 |
| AJ243963.2 | 3 | 551 | CAB60110.1 | |
| AK028225.1 | 3 | 436 | BAC25824.1 | |
| AK031749.1 | 3 | 1152 | BAC27536.1 | |
| BC096448.1 | 3 | 1142 | AAH96448.1 |

Genomic Map