Assay Details
Target Gene Details
Entrez Gene ID: | 60596 |
Gene Name: | guanylate cyclase 1, soluble, alpha 3 |
Gene Aliases: |
1200016O07Rik, sGC-alpha1 |
Location: |
Chr.3:82092427-82145877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy1a3 | NM_021896.5 | NP_068696.2 | ||
| AF297082.1 | AAG17446.1 | |||
| AK004815.1 | BAB23586.1 | |||
| AK028558.1 | BAC26009.1 | |||
| AK031305.1 | ||||
| BC057327.1 | AAH57327.1 | |||
| BC094310.1 | AAH94310.1 |
More Information
Set Membership: |
|

Genomic Map