Assay Details
Target Gene Details
Entrez Gene ID: | 15493 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 |
Gene Aliases: |
- |
Location: |
Chr.3:98711107-98724594 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd3b2 |

Genomic Map