Assay Details
Target Gene Details
Entrez Gene ID: | 13038 |
Gene Name: | cathepsin K |
Gene Aliases: |
AI323530, MMS10-Q, Ms10q, catK |
Location: |
Chr.3:95499210-95509387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsk | NM_007802.4 | NP_031828.2 | ||
| AK003425.1 | BAB22783.1 | |||
| AK132648.1 | BAE21279.1 | |||
| BC046320.1 | AAH46320.1 | |||
| X94444.1 | CAA64218.1 |
More Information
Set Membership: |
|

Genomic Map