Assay Details
Target Gene Details
Entrez Gene ID: | 23831 |
Gene Name: | carbonic anhydrase 14 |
Gene Aliases: |
AW536446, Ca14 |
Location: |
Chr.3:95897768-95905118 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car14 | NM_011797.2 | 7 | 904 | NP_035927.1 |
| AB005450.1 | 7 | 904 | BAA78709.1 | |
| AK012316.1 | 4 | 281 | ||
| AK034693.1 | 1 | 178 | ||
| AK157764.1 | 7 | 904 | ||
| BC046995.1 | 7 | 886 | AAH46995.1 |

Genomic Map