Assay Details
Target Gene Details
Entrez Gene ID: | 12346 |
Gene Name: | carbonic anhydrase 1 |
Gene Aliases: |
AW555628, Ca1, Car-1 |
Location: |
Chr.3:14766214-14808365 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car1 | NM_001083957.1 | 6 | 629 | NP_001077426.1 |
| NM_009799.4 | 7 | 641 | NP_033929.2 | |
| AK003066.1 | 7 | 642 | BAB22544.1 | |
| AK033719.1 | 6 | 631 | ||
| AK146372.1 | 7 | 642 | BAE27121.1 | |
| AK162331.1 | 6 | 629 | BAE36857.1 | |
| AK167969.1 | 7 | 641 | BAE39964.1 | |
| AK172076.1 | 7 | 642 | BAE42813.1 | |
| BC011223.1 | 6 | 613 | AAH11223.1 | |
| BC110681.1 | 6 | 611 | AAI10682.1 | |
| BC132432.1 | 7 | 597 | ||
| BC132434.1 | 7 | 597 | ||
| M32452.1 | 6 | 598 | AAA37354.1 |

Genomic Map