Assay Details
Target Gene Details
Entrez Gene ID: | 30791 |
Gene Name: | solute carrier family 39 (zinc transporter), member 1 |
Gene Aliases: |
Zip1, Zirtl |
Location: |
Chr.3:90248192-90253613 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a1 | NM_013901.2 | NP_038929.2 | ||
| AK146175.1 | BAE26954.1 | |||
| BC005474.1 | AAH05474.1 |
More Information
Set Membership: |
|

Genomic Map