Assay Details
Target Gene Details
Entrez Gene ID: | 23897 |
Gene Name: | HCLS1 associated X-1 |
Gene Aliases: |
HAX-1, HSP1BP-1, Hs1bp1, SIG-111, Silg111, mHAX-1s |
Location: |
Chr.3:89995446-89998716 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hax1 | NM_001282032.1 | NP_001268961.1 | ||
| NM_011826.4 | NP_035956.1 | |||
| AF023482.1 | AAB81081.1 | |||
| AF023483.1 | ||||
| AF465243.1 | AAL76987.1 | |||
| AK002256.1 | BAB21969.1 | |||
| AK010633.1 | BAB27077.1 | |||
| AK088746.1 | BAC40544.1 | |||
| AK135138.1 | BAE22434.1 | |||
| BC006688.1 | AAH06688.1 | |||
| BC098225.1 | AAH98225.1 | |||
| BC147481.1 |

Genomic Map