Assay Details
Target Gene Details
Entrez Gene ID: | 229503 |
Gene Name: | ribosomal RNA adenine dimethylase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:87922601-87930588 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrnad1 | NM_153562.4 | NP_705790.3 | ||
| AK030684.1 | ||||
| AK035389.1 | BAC29055.1 | |||
| AK082223.1 | ||||
| AK172049.1 | 3 | 1127 | ||
| BC023814.1 | AAH23814.1 |
More Information
Set Membership: |
|

Genomic Map