Assay Details
Target Gene Details
Entrez Gene ID: | 23922 |
Gene Name: | jumping translocation breakpoint |
Gene Aliases: |
Gm622 |
Location: |
Chr.3:90231597-90235840 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jtb | NM_206924.2 | NP_996807.1 | ||
| AB016490.1 | BAA33733.1 | |||
| AK004469.1 | BAB23317.1 | |||
| AK089048.1 | BAC40719.1 | |||
| AK146337.1 | BAE27091.1 | |||
| BC008139.1 | AAH08139.1 | |||
| BG077727.2 |

Genomic Map