Assay Details
Target Gene Details
Entrez Gene ID: | 229600 |
Gene Name: | cDNA sequence BC028528 |
Gene Aliases: |
L259 |
Location: |
Chr.3:95882352-95892949 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC028528 | NM_153513.2 | NP_705733.2 | ||
| AF534879.1 | AAN03482.1 | |||
| BC028528.1 | AAH28528.1 |
Target Gene Details
Entrez Gene ID: | 100042889 |
Gene Name: | RIKEN cDNA C920021L13 gene |
Gene Aliases: |
- |
Location: |
Chr.3:95871522-95889093 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C920021L13Rik | NR_040446.1 | |||
| AK044447.1 | ||||
| AK160363.1 |
More Information
Set Membership: |
|

Genomic Map