Assay Details
Target Gene Details
Entrez Gene ID: | 72413 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 2 |
Gene Aliases: |
2700049B16Rik, 3110031N04Rik |
Location: |
Chr.3:31902132-32200180 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnmb2 | NM_028231.2 | NP_082507.1 | ||
| AK012400.1 | BAB28216.1 | |||
| AK014106.1 | ||||
| BC046227.1 | AAH46227.1 | |||
| BC058957.1 | AAH58957.1 |
More Information
Set Membership: |
|

Genomic Map